Chromone-3-carboxylic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A14422-03
A14422-06
CAAAA14422-03EA
58.14
CAD
CAAAA14422-03
CAAAA14422-06
Chromone-3-carboxylic acid 98%
Chromone-3-carboxylic Acid
Purity: 98%
CAS No.: 39079-62-4
Molecular Formula: C10H6O4
Formula Weight: 190.15
Melting Point: 198-203°
Beilstein Reference: 1074111
Formula:
C₁₀H₆O₄ Melting Pt: 198…203 °C Storage Temperature: Ambient |
MDL Number:
MFCD00017338 CAS Number: 39079-62-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...