Cinnamyl chloride 95% stabilized
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
3-Chloro-1-phenyl-1-propene
Stabilization:
Stabilised with sodium carbonate (Na₂CO₃)
A14408-0B
A14408-18
A14408-30
CAAAA14408-0BEA
1218.18
CAD
CAAAA14408-0B
CAAAA14408-18
CAAAA14408-30
Cinnamyl chloride 95% stabilized
Cinnamyl chloride
Stab. with sodium carbonate
Purity: 95%
CAS No.: 2687-12-9
Molecular Formula: C6H5CH=CHCH2Cl
Formula Weight: 152.62
Storage Temperature: 2°C to 8°C
Flash Point: 79°(174°F)
Refractive Index: 1.5845
Melting Point: 7°
Boiling Point: 86-87°/2mm
Beilstein Reference: 741974
UN No.: 2927
MDL No.: MFCD00000986
Formula:
C₉H₉Cl MW: 152.62 g/mol Boiling Pt: 86…87 °C/2 mm Melting Pt: 7 °C Density: 1.096 Flash Pt: 79 °C(174 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00000986 CAS Number: 2687-12-9 EINECS: 220-246-4 UN: 2922 ADR: "8",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...