2-(Pyridin-4-yl)acetic acid hydrochloride ≥98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A14348-06
A14348-14
CAAAAA14348-03EA
81.7
CAD
CAAAAA14348-03
CAAAA14348-06
CAAAA14348-14
2-(Pyridin-4-yl)acetic acid hydrochloride ≥98%
2-(Pyridin-4-yl)acetic acid hydrochloride
Purity: 98+%
CAS No.: 6622-91-9
Molecular Formula: C7H7NO2.HCl
Formula Weight: 173.60
Storage Temperature: 2°C to 8°C
Odor: Odorless
Melting Point: ca 136° dec.
Beilstein Reference: 3696648
MDL No.: MFCD00012827
Formula:
C₇H₈ClNO₂ Melting Pt: ca 136 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00012827 CAS Number: 6622-91-9 EINECS: 229-576-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...