5,6-Dichloro-2-mercaptobenzimidazole 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5,6-Dichloro-1H-benzimidazole-2-thiol

A14309-03 A14309-06
CAAAA14309-03EA 97.49 CAD
CAAAA14309-03 CAAAA14309-06
5,6-Dichloro-2-mercaptobenzimidazole 98%
5,6-Dichloro-2-mercaptobenzimidazole

Purity: 98% CAS No.: 19462-98-7 Molecular Formula: C7H4Cl2N2S Formula Weight: 219.09 Odor: Odorless Melting Point: ca 350° dec. MDL No.: MFCD00052398

Formula: C₇H₄Cl₂N₂S
MW: 219.09 g/mol
Melting Pt: ∼ 350 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00052398
CAS Number: 19462-98-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR