Bis(3-methyl-1-phenyl-5-pyrazolone) ≥98%

Supplier: Thermo Scientific Chemicals

Synonyms: 3,3-Dimethyl-1,1-diphenyl-1H,1H-[4,4-bipyrazole]-5,5(4H,4H)-dione, 3,3'-Dimethyl-1,1'-diphenyl-4,4'-bi(5-pyrazolone), Bispyrazolone

A14308-06 A14308-14
CAAAA14308-06EA 69.99 CAD
CAAAA14308-06 CAAAA14308-14
Bis(3-methyl-1-phenyl-5-pyrazolone) ≥98%
Bis(3-methyl-1-phenyl-5-pyrazolone)

Purity: 98+% CAS No.: 7477-67-0 Molecular Formula: C20H18N4O2 Formula Weight: 346.39 Melting Point: 300° Beilstein Reference: 572787 MDL No.: MFCD00020765

Formula: C₂₀H₁₈N₄O₂
MW: 346.39 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00020765
CAS Number: 7477-67-0
EINECS: 231-282-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR