Folic acid dihydrate 97%
Supplier: Thermo Scientific Chemicals
A14300-14
A14300-22
CAAAA14300-14EA
123.56
CAD
CAAAA14300-14
CAAAA14300-22
Folic acid dihydrate 97%
Folic acid dihydrate
Vitamin B9. A B-complex vitamin necessary for the body to produce red blood cells
Purity: 97%
CAS No.: 75708-92-8
Molecular Formula: C19H19N7O6.2H2O
Formula Weight: 477.44 (441.40anhy)
Melting Point: ca 250° dec.
Beilstein Reference: 100781
Merck Reference: 14,4221
Optical Rotation: +18° (c=0.5 in 0.1N NaOH)
MDL No.: MFCD00079305
Vitamin B9. A B-complex vitamin necessary for the body to produce red blood cells
Formula:
C₁₉H₁₉N₇O₆·2H₂O Melting Pt: ca 250 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00079305 CAS Number: 75708-92-8 EINECS: 200-419-0 Merck Index: 14,4221 |
Specification Test Results
Optical rotation | +18° (c=0.5 in 0.1N NaOH) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...