2,6-Lutidine-N-oxide 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A14299-09
A14299-18
A14299-30
CAAAA14299-09EA
132.25
CAD
CAAAA14299-09
CAAAA14299-18
CAAAA14299-30
2,6-Lutidine-N-oxide 98%
2,6-Lutidine-N-oxide
Purity: 98%
CAS No.: 1073-23-0
Molecular Formula: C7H9NO
Formula Weight: 123.16
Refractive Index: 1.5685
Boiling Point: 108-111°/10mm
Beilstein Reference: 110551
MDL No.: MFCD00039715
Formula:
C₇H₉NO Boiling Pt: 108…111 °C/10 mm Density: 1.102 Storage Temperature: Ambient |
MDL Number:
MFCD00039715 CAS Number: 1073-23-0 EINECS: 214-025-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...