ɑ-Bromocinnamaldehyde 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Bromo-3-phenyl-2-propenal, 2-Bromo-3-phenylacrylaldehyde, Alpha-Bromocinnamaldehyde

A14230-14 A14230-22
CAAAA14230-14EA 75.68 CAD
CAAAA14230-14 CAAAA14230-22 CAAAA14230-36
ɑ-Bromocinnamaldehyde 98%
alpha-Bromocinnamaldehyde

Purity: 98% CAS No.: 5443-49-2 Molecular Formula: C9H7BrO Formula Weight: 211.06 Melting Point: 68-72° MDL No.: MFCD00006965

Formula: C₉H₇BrO
Melting Pt: 68…72 °C
Storage Temperature: Ambient
MDL Number: MFCD00006965
CAS Number: 5443-49-2
EINECS: 226-637-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR