3-Ethoxy-2-cyclohexen-1-one 99%
Supplier: Thermo Scientific Chemicals
A14224-09
A14224-18
A14224-30
CAAAA14224-09EA
74.89
CAD
CAAAA14224-09
CAAAA14224-18
CAAAA14224-30
3-Ethoxy-2-cyclohexen-1-one 99%
3-Ethoxy-2-cyclohexen-1-one
Purity: 99%
CAS No.: 5323-87-5
Molecular Formula: C8H12O2
Formula Weight: 140.18
Storage Temperature: -30°C to -10°C
Flash Point: 107°(224°F)
Refractive Index: 1.5060
Boiling Point: 76-78°/1mm
Beilstein Reference: 2042049
MDL No.: MFCD00001580
Formula:
C₈H₁₂O₂ Boiling Pt: 76…78 °C/1 mm Density: 1.04 Flash Pt: 107 °C(224 °F) Storage Temperature: Freezer |
MDL Number:
MFCD00001580 CAS Number: 5323-87-5 EINECS: 226-190-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...