Tetraethyl ethylenebisphosphonate 98%

Supplier: Thermo Scientific Chemicals
Warning

A14206-03 A14206-06 A14206-14
CAAAA14206-03EA 41.87 CAD
CAAAA14206-03 CAAAA14206-06 CAAAA14206-14
Tetraethyl ethylenebisphosphonate 98%
Tetraethyl ethylenebisphosphonate

Purity: 98% CAS No.: 995-32-4 Molecular Formula: C10H24O6P2 Formula Weight: 302.25 Flash Point: 110°(230°F) Refractive Index: 1.4410 Boiling Point: 153-154°/0.9mm Beilstein Reference: 1800096 MDL No.: MFCD00015133

Formula: C₁₀H₂₄O₆P₂
Boiling Pt: 153…154 °C/0.9 mm
Density: 1,146
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00015133
CAS Number: 995-32-4
EINECS: 213-625-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR