1,3-Dihydroxyacetone dimer 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A14189-0B
A14189-18
A14189-30
CAAAA14189-0BEA
559.32
CAD
CAAAA14189-0B
CAAAA14189-18
CAAAA14189-30
1,3-Dihydroxyacetone dimer 97%
1,3-Dihydroxyacetone dimer
Purity: 97%
CAS No.: 62147-49-3
Molecular Formula: C6H12O6
Formula Weight: 180.16
Storage Temperature: 2°C to 8°C
Melting Point: ca 75-82°
Beilstein Reference: 112910
Merck Reference: 14,3182
MDL No.: MFCD00051019
Formula:
C₆H₁₂O₆ Melting Pt: ca 75…82 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00051019 CAS Number: 62147-49-3 EINECS: 612-947-2 Merck Index: 14,3182 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...