2-Phenyl-2-butenal mixture E/Z 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A14185-06
A14185-14
CAAAA14185-06EA
77.1
CAD
CAAAA14185-06
CAAAA14185-14
2-Phenyl-2-butenal mixture E/Z 97%
2-Phenyl-2-butenal
Purity: 97%
CAS No.: 4411-89-6
Molecular Formula: C10H10O
Formula Weight: 146.19
Refractive Index: 1.5610
Boiling Point: 115°/15mm
MDL No.: MFCD00053158
Formula:
C₁₀H₁₀O Boiling Pt: 115 °C/15 mm Storage Temperature: Ambient |
MDL Number:
MFCD00053158 CAS Number: 4411-89-6 EINECS: 224-567-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...