N-Phenylglycine 97%
Supplier: Thermo Scientific Chemicals
Synonyms:
2-Anilinoacetic acid, 2-(Phenylamino)acetic acid
A14182-09
A14182-18
CAAAA14182-09EA
59.41
CAD
CAAAA14182-09
CAAAA14182-18
N-Phenylglycine 97%
N-Phenylglycine
Applications: Pharmaceutical intermediate.
Purity: 97%
CAS No.: 103-01-5
Molecular Formula: (C6H5)NHCH2CO2H
Formula Weight: 151.17
Melting Point: 117-119°
Beilstein Reference: 509838
Merck Reference: 14,7292
MDL No.: MFCD00014009
Formula:
C₈H₉NO₂ MW: 151.17 g/mol Melting Pt: 117…119 °C Storage Temperature: Ambient |
MDL Number:
MFCD00014009 CAS Number: 103-01-5 EINECS: 203-070-2 Merck Index: 14,7292 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...