L-Serine ethyl ester hydrochloride 99%
Supplier: Thermo Scientific Chemicals
A14174-06
A14174-14
CAAAA14174-06EA
82.48
CAD
CAAAA14174-06
CAAAA14174-14
CAAAA14174-22
L-Serine ethyl ester hydrochloride 99%
L-Serine ethyl ester hydrochloride
Purity: 99%
CAS No.: 26348-61-8
Molecular Formula: C5H11NO3.HCl
Formula Weight: 169.61
Storage Temperature: -30°C to -10°C
Melting Point: 130-132°
Beilstein Reference: 3562346
Optical Rotation: -4.4° (c=4 in water)
MDL No.: MFCD00012594
Formula:
C₅H₁₂ClNO₃ Melting Pt: 130…132 °C Storage Temperature: Freezer |
MDL Number:
MFCD00012594 CAS Number: 26348-61-8 EINECS: 247-624-1 |
Specification Test Results
Optical rotation | -4.4° (c=4 in water) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...