2-(Hydroxymethyl)-1,4-benzodioxane 98%

Supplier: Thermo Scientific Chemicals

A14170-06 A14170-14
CAAAA14170-06EA 43.29 CAD
CAAAA14170-06 CAAAA14170-14
2-(Hydroxymethyl)-1,4-benzodioxane 98%
2-(Hydroxymethyl)-1,4-benzodioxane

Purity: 98% CAS No.: 3663-82-9 Molecular Formula: C9H10O3 Formula Weight: 166.18 Melting Point: 87-91° Beilstein Reference: 83736

Formula: C₉H₁₀O₃
Melting Pt: 87…91 °C
Storage Temperature: Ambient
MDL Number: MFCD00006822
CAS Number: 3663-82-9
EINECS: 222-921-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR