6-Aza-2-thiothymine 98%

Supplier: Thermo Scientific Chemicals

Synonyms: 6-Methyl-3-thioxo-3,4-dihydro-1,2,4-triazin-5(2H)-one

A14167-06 A14167-14 A14167-22
CAAAA14167-06EA 143.46 CAD
CAAAA14167-06 CAAAA14167-14 CAAAA14167-22
6-Aza-2-thiothymine 98%
6-Aza-2-thiothymine

Purity: 98% CAS No.: 615-76-9 Molecular Formula: C4H5N3OS Formula Weight: 143.17 Melting Point: 218-221° Beilstein Reference: 4861 MDL No.: MFCD00006458

Formula: C₄H₅N₃OS
Melting Pt: 218…221 °C
Storage Temperature: Ambient
MDL Number: MFCD00006458
CAS Number: 615-76-9
EINECS: 210-445-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR