Ethyl phenoxyacetate 99%

Supplier: Thermo Scientific Chemicals

A14156-30 A14156-09 A14156-18
CAAAAA14156-30EA 707.84 CAD
CAAAAA14156-30 CAAAA14156-09 CAAAA14156-18
Ethyl phenoxyacetate 99%
Ethyl phenoxyacetate

Purity: 99% CAS No.: 2555-49-9 Molecular Formula: C6H5OCH2CO2CH2CH3 Formula Weight: 180.20 Flash Point: 110°(230°F) Refractive Index: 1.5055 Boiling Point: 251-253° Beilstein Reference: 1871712 MDL No.: MFCD00026895

Formula: C₁₀H₁₂O₃
Boiling Pt: 251…253 °C
Density: 1,108
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00026895
CAS Number: 2555-49-9
EINECS: 219-867-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR