4-Fluoro-ɑ-methylstyrene 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Fluoro-4-(1-methylethenyl)benzene, 2-(4-Fluorophenyl)-1-propene

A14147-09 A14147-18 A14147-30
CAAAA14147-09EA 178.54 CAD
CAAAA14147-09 CAAAA14147-18 CAAAA14147-30
4-Fluoro-ɑ-methylstyrene 95%
4-Fluoro-alpha-methylstyrene

Purity: 95% CAS No.: 350-40-3 Molecular Formula: C9H9F Formula Weight: 136.17 Storage Temperature: 2°C to 8°C Flash Point: 53°(127°F) Refractive Index: 1.5110 Boiling Point: 183° Beilstein Reference: 2039696 UN No.: 1993 MDL No.: MFCD00042297

Formula: C₉H₉F
MW: 136.17 g/mol
Boiling Pt: 183 °C
Density: 1.009
Flash Pt: 53 °C (127 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00042297
CAS Number: 350-40-3
EINECS: 206-501-2
UN: 1993
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR