(1R)-(+)-2,10,Camphorsultam 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A14133-03
A14133-06
CAAAA14133-03EA
131.61
CAD
CAAAA14133-03
CAAAA14133-06
(1R)-(+)-2,10,Camphorsultam 99%
(1R)-(+)-2,10,Camphorsultam
Fieser: 13,62 14,71 17,65
Purity: 99%
CAS No.: 108448-77-7
Molecular Formula: C10H17NO2S
Formula Weight: 215.32
Melting Point: 179-186°
Beilstein Reference: 4675755
Optical Rotation: +33° (c=1 in ethanol)
MDL No.: MFCD00151762
Formula:
C₁₀H₁₇NO₂S Melting Pt: 179…186 °C Storage Temperature: Ambient |
MDL Number:
MFCD00151762 CAS Number: 108448-77-7 |
Specification Test Results
Optical rotation | +33° (c=1 in ethanol) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...