4-tert-Butylthiazol-2-ylamine 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A14115-03
A14115-06
A14115-14
CAAAA14115-03EA
49.61
CAD
CAAAA14115-03
CAAAA14115-06
CAAAA14115-14
4-tert-Butylthiazol-2-ylamine 98%
4-tert-Butylthiazol-2-ylamine
Purity: 98%
CAS No.: 74370-93-7
Molecular Formula: C7H12N2S
Formula Weight: 156.26
Melting Point: 98-103°
Beilstein Reference: 114270
MDL No.: MFCD00051644
Formula:
C₇H₁₂N₂S Melting Pt: 98…103 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051644 CAS Number: 74370-93-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...