Benzo[b]thiophene-1,1-dioxide 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Benzothiophene 1,1-Dioxide

A14063-06 A14063-14
CAAAA14063-06EA 164.32 CAD
CAAAA14063-06 CAAAA14063-14
Benzo[b]thiophene-1,1-dioxide 98%
Benzo[b]thiophene-1,1-dioxide

Purity: 98% CAS No.: 825-44-5 Molecular Formula: C8H6O2S Formula Weight: 166.20 Melting Point: 137-138° Beilstein Reference: 116442 MDL No.: MFCD00052178

Formula: C₈H₆O₂S
MW: 166.20 g/mol
Melting Pt: 137…138 °C
Storage Temperature: Ambient
MDL Number: MFCD00052178
CAS Number: 825-44-5
EINECS: 212-544-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR