Tropylium tetrafluoroborate 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Tropyliumtetrafluoroborate, Cyclohepta-2,4,6-trienylium tetrafluoroborate

A14045-06 A14045-14 A14045-22
CAAAA14045-06EA 328.64 CAD
CAAAA14045-06 CAAAA14045-14 CAAAA14045-22
Tropylium tetrafluoroborate 97%
Tropylium tetrafluoroborate

Fieser: 1,1261


Purity: 97% CAS No.: 27081-10-3 Molecular Formula: C7H7BF4 Formula Weight: 177.94 Storage Temperature: 2°C to 8°C Melting Point: ca 240° dec. Beilstein Reference: 3574512 UN No.: 1759 MDL No.: MFCD00013402

Formula: C₇H₇BF₄
Melting Pt: ca 240 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00013402
CAS Number: 27081-10-3
EINECS: 248-214-5
UN: 3261
ADR: "8",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR