Phenylpyruvic acid sodium salt monohydrate 98%

Supplier: Thermo Scientific Chemicals

Synonyms: 2-Oxo-3-phenylpropionic acid sodium salt monohydrate, Sodium phenylpyruvate monohydrate, Sodium 2-oxo-3-phenylpropanoate monohydrate

A14027-18 A14027-03 A14027-06 A14027-14
CAAAAA14027-18EA 505.6 CAD
CAAAAA14027-18 CAAAA14027-03 CAAAA14027-06 CAAAA14027-14
Phenylpyruvic acid sodium salt monohydrate 98%
Phenylpyruvic acid sodium salt monohydrate

Purity: 98% CAS No.: 122049-54-1 Molecular Formula: C9H7NaO3.H2O Formula Weight: 204.16 (186.14anhy) Storage Temperature: 2°C to 8°C Melting Point: 300° Beilstein Reference: 4770978 MDL No.: MFCD00149109

Formula: C₉H₇NaO₃·H₂O
MW: 204.157 g/mol
Melting Pt: >300 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00149109
CAS Number: 122049-54-1
EINECS: 204-053-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR