Monoacetin mixture of isomers cont. varying amounts of diacetate, tech.

Supplier: Thermo Scientific Chemicals

Synonyms: Glycerol Monoacetate, 1,2,3-Propanetriol, monoacetate

A14025-0B A14025-30
CAAAA14025-0BEA 560.9 CAD
CAAAA14025-0B CAAAA14025-30
Monoacetin mixture of isomers cont. varying amounts of diacetate, tech.
Monoacetin

CAS No.: 26446-35-5 Molecular Formula: C5H10O4 Formula Weight: 134.13 Flash Point: 145°(293°F) Refractive Index: 1.4500 Boiling Point: 258° MDL No.: MFCD00036185

Formula: C₅H₁₀O₄
MW: 134.13 g/mol
Boiling Pt: 258 °C
Density: 1.21
Flash Pt: 145 °C(293 °F)
Storage Temperature: Ambient
MDL Number: MFCD00036185
CAS Number: 26446-35-5
EINECS: 247-704-6
Merck Index: 14,6242

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR