Isosorbide 98%
Supplier: Thermo Scientific Chemicals
A13989-0B
A13989-18
A13989-30
CAAAA13989-0BEA
630.42
CAD
CAAAA13989-0B
CAAAA13989-18
CAAAA13989-30
CAAAAA13989-D9
Isosorbide 98%
Isosorbide
Purity: 98%
CAS No.: 652-67-5
Molecular Formula: C6H10O4
Formula Weight: 146.14
Odor: Odorless
Melting Point: 56-64°
Beilstein Reference: 80510
Merck Reference: 14,5224
Optical Rotation: +45° (c=3 in water)
MDL No.: MFCD00064827
Formula:
C₆H₁₀O₄ Melting Pt: 56…64 °C Storage Temperature: Ambient |
MDL Number:
MFCD00064827 CAS Number: 652-67-5 EINECS: 211-492-3 Merck Index: 14,5224 |
Specification Test Results
Optical rotation | +45° (c=3 in water) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...