1,2-Diamino-4-chloro-5-fluorobenzene 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Chloro-5-fluorobenzene-1,2-diamine

A13953-03 A13953-06
CAAAA13953-03EA 85.64 CAD
CAAAA13953-03 CAAAA13953-06 CAAAA13953-14
1,2-Diamino-4-chloro-5-fluorobenzene 98%
1,2-Diamino-4-chloro-5-fluorobenzene

Purity: 98% CAS No.: 139512-70-2 Molecular Formula: C6H6ClFN2 Formula Weight: 160.58 Melting Point: 110-114° UN No.: 2811 MDL No.: MFCD00042485

Formula: C₆H₆ClFN₂
MW: 160.58 g/mol
Melting Pt: 110…114 °C
Storage Temperature: Ambient
MDL Number: MFCD00042485
CAS Number: 139512-70-2
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR