3-Amino-4-methoxybenzamide 98%

Supplier: Thermo Scientific Chemicals
Warning

A13942-06 A13942-14
CAAAA13942-06EA 38.87 CAD
CAAAA13942-06 CAAAA13942-14 CAAAA13942-22
3-Amino-4-methoxybenzamide 98%
3-Amino-4-methoxybenzamide

Purity: 98% CAS No.: 17481-27-5 Molecular Formula: C8H10N2O2 Formula Weight: 166.18 Melting Point: 156-160° MDL No.: MFCD00017132

Formula: C₈H₁₀N₂O₂
Melting Pt: 156…160 °C
Storage Temperature: Ambient
MDL Number: MFCD00017132
CAS Number: 17481-27-5
EINECS: 241-497-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR