4-Hydroxy-3,5-dimethoxybenzyl alcohol 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A13888-03
A13888-06
A13888-14
CAAAAA13888-03EA
56.25
CAD
CAAAAA13888-03
CAAAA13888-06
CAAAA13888-14
4-Hydroxy-3,5-dimethoxybenzyl alcohol 97%
4-Hydroxy-3,5-dimethoxybenzyl alcohol
Purity: 97%
CAS No.: 530-56-3
Molecular Formula: C9H12O4
Formula Weight: 184.19
Melting Point: 129-132°
Beilstein Reference: 1873975
MDL No.: MFCD00016871
Formula:
C₉H₁₂O₄ Melting Pt: 129…132 °C Storage Temperature: Ambient |
MDL Number:
MFCD00016871 CAS Number: 530-56-3 EINECS: 208-485-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...