N,N'-Bis(salicylidene)ethylenediamine 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Ethylenebis(salicylimine), 2,2'-((Ethane-1,2-diylbis(azanylylidene))bis(methanylylidene))diphenol, SALEN

A13885-14 A13885-22 A13885-36
CAAAA13885-14EA 119.29 CAD
CAAAA13885-14 CAAAA13885-22 CAAAA13885-36
N,N'-Bis(salicylidene)ethylenediamine 98%
N,N'-Bis(salicylidene)ethylenediamine

Purity: 98% CAS No.: 94-93-9 Molecular Formula: C16H16N2O2 Formula Weight: 268.32 Melting Point: 129-131° Beilstein Reference: 535296 Merck Reference: 14,8322 MDL No.: MFCD00002244

Formula: C₁₆H₁₆N₂O₂
MW: 268.31 g/mol
Melting Pt: 129…131 °C
Storage Temperature: Ambient
MDL Number: MFCD00002244
CAS Number: 94-93-9
EINECS: 202-376-3
Merck Index: 14,8322

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR