Triphenylmethylamine 98%

Supplier: Thermo Scientific Chemicals
Warning

A13864-03 A13864-06 A13864-14 A13864-22
CAAAAA13864-03EA 43.92 CAD
CAAAAA13864-03 CAAAA13864-06 CAAAA13864-14 CAAAA13864-22
Triphenylmethylamine 98%
Triphenylmethylamine

Purity: 98% CAS No.: 5824-40-8 Molecular Formula: (C6H5)3CNH2 Formula Weight: 259.35 Melting Point: 100-104° Boiling Point: 223°/14mm Beilstein Reference: 2113674 MDL No.: MFCD00008047

Formula: C₁₉H₁₇N
Boiling Pt: 223 °C/14 mm
Melting Pt: 100…104 °C
Storage Temperature: Ambient
MDL Number: MFCD00008047
CAS Number: 5824-40-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR