N-Benzylaminoacetaldehyde diethyl acetal 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A13841-06
A13841-14
CAAAA13841-03EA
64.48
CAD
CAAAA13841-03
CAAAA13841-06
CAAAA13841-14
N-Benzylaminoacetaldehyde diethyl acetal 96%
N-Benzylaminoacetaldehyde diethyl acetal
Purity: 96%
CAS No.: 61190-10-1
Molecular Formula: C6H5CH2NHCH2CH(OCH2CH3)2
Formula Weight: 223.32
Refractive Index: 1.4890
Boiling Point: 115-117°/1mm
MDL No.: MFCD00051599
Formula:
C₁₃H₂₁NO₂ Boiling Pt: 115…117 °C/1 mm Storage Temperature: Ambient |
MDL Number:
MFCD00051599 CAS Number: 61190-10-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...