10-Undecynoic acid 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A13815-06
A13815-14
CAAAA13815-06EA
172.22
CAD
CAAAA13815-06
CAAAA13815-14
10-Undecynoic acid 96%
10-Undecynoic acid
Purity: 96%
CAS No.: 2777-65-3
Molecular Formula: HC≡C(CH2)8CO2H
Formula Weight: 182.26
Flash Point: 161°(321°F)
Odor: Odorless
Boiling Point: 174-176°/4mm
Beilstein Reference: 1704918
MDL No.: MFCD00014389
Formula:
C₁₁H₁₈O₂ Boiling Pt: 174…176 °C/4 mm Melting Pt: 40…43 °C Flash Pt: 161 °C(321 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00014389 CAS Number: 2777-65-3 EINECS: 220-471-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...