Ethyl-3,4,5-trimethylpyrrole-2-carboxylate 98%
Supplier: Thermo Scientific Chemicals
A13814-03
A13814-06
CAAAA13814-03EA
63.2
CAD
CAAAA13814-03
CAAAA13814-06
Ethyl-3,4,5-trimethylpyrrole-2-carboxylate 98%
Ethyl 3,4,5-trimethylpyrrole-2-carboxylate
Purity: 98%
CAS No.: 2199-46-4
Molecular Formula: C10H15NO2
Formula Weight: 181.24
Melting Point: 124-128°
Beilstein Reference: 132613
Formula:
C₁₀H₁₅NO₂ Melting Pt: 124…128 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051948 CAS Number: 2199-46-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...