Ethyl acetopyruvate 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Ethyl 2,4-dioxopentanoate, Ethyl 2,4-dioxovalerate

A13808-06 A13808-14 A13808-22
CAAAA13808-06EA 87.06 CAD
CAAAA13808-06 CAAAA13808-14 CAAAA13808-22
Ethyl acetopyruvate 98%
Ethyl acetopyruvate

Purity: 98% CAS No.: 615-79-2 Molecular Formula: C7H10O4 Formula Weight: 158.15 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Refractive Index: 1.4730 Melting Point: 16-18° Boiling Point: 108-109°/12mm Beilstein Reference: 607062 MDL No.: MFCD00009124

Formula: C₇H₁₀O₄
Boiling Pt: 108…109 °C/12 mm
Melting Pt: 16…18 °C
Density: 1.126
Flash Pt: >110 °C(230 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00009124
CAS Number: 615-79-2
EINECS: 210-447-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR