4-(Dimethylamino)benzhydrazide 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A13723-09
A13723-18
CAAAA13723-09EA
96.7
CAD
CAAAA13723-09
CAAAA13723-18
CAAAA13723-30
4-(Dimethylamino)benzhydrazide 98+%
4-(Dimethylamino)benzhydrazide
Purity: 98+%
CAS No.: 19353-92-5
Molecular Formula: C9H13N3O
Formula Weight: 179.22
Melting Point: 174-175°
Beilstein Reference: 2210856
Formula:
C₉H₁₃N₃O Melting Pt: 174…175 °C Storage Temperature: Ambient |
MDL Number:
MFCD00014761 CAS Number: 19353-92-5 EINECS: 242-985-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...