Crotonoyl chloride 90% remainder mainly cis-isomer, tech., Technical Grade
Supplier: Thermo Scientific Chemicals
|
Danger
|
A13710-14
A13710-22
A13710-36
CAAAA13710-14EA
66.04
CAD
CAAAA13710-14
CAAAA13710-22
CAAAA13710-36
Crotonoyl chloride 90% remainder mainly cis-isomer, tech., Technical Grade
Crotonoyl chloride
Purity: 90%
CAS No.: 625-35-4
Molecular Formula: C4H5ClO
Formula Weight: 104.54
Storage Temperature: 2°C to 8°C
Flash Point: 35°(95°F)
Refractive Index: 1.4600
Odor: Acrid
Boiling Point: 121-123°
Beilstein Reference: 506118
UN No.: 2920
MDL No.: MFCD00000734
Formula:
C₄H₅ClO Boiling Pt: 121…123 °C Density: 1.091 Flash Pt: 35 °C(95 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00000734 CAS Number: 10487-71-5 EINECS: 210-889-9 UN: 2920 ADR: "8",II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...