2-(Phenylthio)thiophene 97+%

Supplier: Thermo Scientific Chemicals

A13708-03 A13708-06
CAAAA13708-03EA 114.87 CAD
CAAAA13708-03 CAAAA13708-06
2-(Phenylthio)thiophene 97+%
2-(Phenylthio)thiophene

Purity: 97+% CAS No.: 16718-12-0 Molecular Formula: C10H8S2 Formula Weight: 192.30 Boiling Point: 109-110°/0.4mm MDL No.: MFCD00052141

Formula: C₁₀H₈S₂
MW: 192.30 g/mol
Boiling Pt: 109…110 °C/0.4 mm
Storage Temperature: Ambient
MDL Number: MFCD00052141
CAS Number: 16718-12-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR