4-Methoxycoumarin 98%
Supplier: Thermo Scientific Chemicals
A13670-06
A13670-14
CAAAAA13670-22EA
1582.37
CAD
CAAAAA13670-22
CAAAA13670-06
CAAAA13670-14
4-Methoxycoumarin 98%
4-Methoxycoumarin
Purity: 98%
CAS No.: 20280-81-3
Molecular Formula: C10H8O3
Formula Weight: 176.17
Melting Point: 122-125°
Beilstein Reference: 145960
MDL No.: MFCD00017349
Formula:
C₁₀H₈O₃ MW: 176.17 g/mol Melting Pt: 122…125 °C Storage Temperature: Ambient |
MDL Number:
MFCD00017349 CAS Number: 20280-81-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...