Diethyl phenylthiomethylphosphonate 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A13660-09
A13660-18
CAAAAA13660-09EA
114.55
CAD
CAAAAA13660-09
CAAAAA13660-18
CAAAAA13660-30
Diethyl phenylthiomethylphosphonate 96%
Diethyl phenylthiomethylphosphonate
Purity: 96%
CAS No.: 38066-16-9
Molecular Formula: C11H17O3PS
Formula Weight: 260.29
Refractive Index: 1.5340
Boiling Point: 146-148°/0.5mm
Beilstein Reference: 1246215
MDL No.: MFCD00015130
Formula:
C₁₁H₁₇O₃PS Boiling Pt: 146…148 °C/0.5 mm Density: 1.174 Storage Temperature: Ambient |
MDL Number:
MFCD00015130 CAS Number: 38066-16-9 EINECS: 253-770-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...