(±)-Aspartic acid 98+%
Supplier: Thermo Scientific Chemicals
A13646-0E
A13646-22
A13646-36
CAAAA13646-0EEA
317.58
CAD
CAAAA13646-0E
CAAAA13646-22
CAAAA13646-36
(±)-Aspartic acid 98+%
(±)-Aspartic acid
Purity: 98+%
CAS No.: 617-45-8
Molecular Formula: C4H7NO4
Formula Weight: 133.10
Melting Point:
Formula:
C₄H₇NO₄ Melting Pt: >300 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00063083 CAS Number: 617-45-8 EINECS: 210-513-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...