2-Phenyl-2-pentenal mixture E/Z ≥90%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A13635-06
A13635-14
CAAAAA13635-22EA
951.14
CAD
CAAAAA13635-22
CAAAA13635-06
CAAAA13635-14
2-Phenyl-2-pentenal mixture E/Z ≥90%
2-Phenyl-2-pentenal
Purity: 90+%
CAS No.: 3491-63-2
Molecular Formula: C11H12O
Formula Weight: 160.22
Storage Temperature: 2°C to 8°C
Flash Point:
Formula:
C₁₁H₁₂O MW: 160.22 g/mol Boiling Pt: 124…126 °C/14 mm Density: 1.005 Flash Pt: >100 °C (212 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00051835 CAS Number: 3491-63-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...