L-Phenylalanine ethyl ester hydrochloride 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Ethyl L-Phenylalaninate HCl, H-Phe-OEt·HCl, Ethyl L-phenylalaninate hydrochloride

A13618-06 A13618-14 A13618-22
CAAAA13618-06EA 51.98 CAD
CAAAA13618-06 CAAAA13618-14 CAAAA13618-22
L-Phenylalanine ethyl ester hydrochloride 98%
L-Phenylalanine ethyl ester hydrochloride

Purity: 98% CAS No.: 3182-93-2 Molecular Formula: C11H15NO2.HCl Formula Weight: 229.71 Melting Point: 153-157° Beilstein Reference: 3657823 Optical Rotation: +36° (c=5 in ethanol) MDL No.: MFCD00012507

Formula: C₁₁H₁₆ClNO₂
Melting Pt: 153…157 °C
Storage Temperature: Ambient
MDL Number: MFCD00012507
CAS Number: 3182-93-2
EINECS: 221-673-9

Order Now

Specification Test Results

Optical rotation +36° (c=5 in ethanol)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR