Ethyl-3-(trifluoromethyl)pyrazole-4-carboxylate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Ethyl 3-(trifluoromethyl)pyrazole-4-carboxylate, 3-(Trifluoromethyl)pyrazole-4-carboxylic Acid Ethyl Ester, Ethyl 3-trifluoromethylpyrazole-4-carboxylate
A13609-14
A13609-06
CAAAAA13609-14EA
442.4
CAD
CAAAAA13609-14
CAAAA13609-03
CAAAA13609-06
Ethyl-3-(trifluoromethyl)pyrazole-4-carboxylate 97%
Ethyl-3-(trifluoromethyl)pyrazole-4-carboxylate
Purity: 97%
CAS No.: 155377-19-8
Molecular Formula: C7H7F3N2O2
Formula Weight: 208.14
Melting Point: 142-146°
MDL No.: MFCD00052083
Formula:
C₇H₇F₃N₂O₂ MW: 208.14 g/mol Melting Pt: 142…146 °C Storage Temperature: Ambient |
MDL Number:
MFCD00052083 CAS Number: 155377-19-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...