3',4'-(Methylenedioxy)acetophenone 98%
Supplier: Thermo Scientific Chemicals
A13597-09
A13597-18
A13597-30
CAAAA13597-09EA
70.15
CAD
CAAAA13597-09
CAAAA13597-18
CAAAA13597-30
3',4'-(Methylenedioxy)acetophenone 98%
3',4'-(Methylenedioxy)acetophenone
Purity: 98%
CAS No.: 3162-29-6
Molecular Formula: C9H8O3
Formula Weight: 164.16
Melting Point: 85-89°
Boiling Point: 152-155°/15mm
MDL No.: MFCD00005831
Formula:
C₉H₈O₃ Boiling Pt: 152…155 °C/15 mm Melting Pt: 85…89 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005831 CAS Number: 3162-29-6 EINECS: 221-613-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...