Ammonium zinc sulfate hydrate

Supplier: Thermo Scientific Chemicals
Warning

A13584-0B A13584-30
CAAAAA13584-0BEA 161.16 CAD
CAAAAA13584-0B CAAAAA13584-0I CAAAAA13584-30
Ammonium zinc sulfate hydrate
Ammonium zinc sulfate hydrate

CAS No.: 77998-33-5 Molecular Formula: (NH4)2Zn(SO4)2.xH2O Formula Weight: 293.57(anhy) UN No.: 3077 MDL No.: MFCD00156122

Formula: (NH₄)₂Zn(SO₄)₂·xH₂O
Density: 1.931
Storage Temperature: Ambient
MDL Number: MFCD00156122
CAS Number: 77998-33-5
UN: 3077
ADR: "9",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR