N-Carbobenzyloxy-L-valine 99%
Supplier: Thermo Scientific Chemicals
A13583-14
A13583-22
CAAAA13583-06EA
56.37
CAD
CAAAA13583-06
CAAAA13583-14
CAAAA13583-22
N-Carbobenzyloxy-L-valine 99%
N-Carbobenzyloxy-L-valine
Purity: 99%
CAS No.: 1149-26-4
Molecular Formula: C13H17NO4
Formula Weight: 251.28
Melting Point: 60°
Beilstein Reference: 2056617
MDL No.: MFCD00008922
Formula:
C13H17NO4 Melting Pt: 60 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008922 CAS Number: 1149-26-4 EINECS: 214-562-1 |
Specification Test Results
Optical rotation | -4.0° (c=2 in acetic acid) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...