Methyl-2-(aminosulfonyl)benzoate 98%

Supplier: Thermo Scientific Chemicals
Warning

A13553-14 A13553-22
CAAAA13553-06EA 46.09 CAD
CAAAA13553-06 CAAAA13553-14 CAAAA13553-22
Methyl-2-(aminosulfonyl)benzoate 98%
Methyl-2-(aminosulfonyl)benzoate

Purity: 98% CAS No.: 57683-71-3 Molecular Formula: C8H9NO4S Formula Weight: 215.23 Melting Point: 123-125° MDL No.: MFCD00009808

Formula: C₈H₉NO₄S
Melting Pt: 123…125 °C
Storage Temperature: Ambient
MDL Number: MFCD00009808
CAS Number: 57683-71-3
EINECS: 260-903-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR