1,4-Bis(2-methylstyryl)benzene 99%
Supplier: Thermo Scientific Chemicals
Synonyms:
Bis-MSB, 1,4-Di(2-methylstyryl)benzene, 1-Methyl-2-(2-(4-[2-(2-methylphenyl)ethenyl]phenyl)ethenyl)benzene, p-Bis(o-Methylstyryl) Benzene
A13534-06
A13534-14
CAAAA13534-06EA
119.29
CAD
CAAAA13534-06
CAAAA13534-14
1,4-Bis(2-methylstyryl)benzene 99%
1,4-Bis(2-methylstyryl)benzene
Purity: 99%
CAS No.: 13280-61-0
Molecular Formula: C24H22
Formula Weight: 310.44
Odor: Odorless
Melting Point: 179-184°
Beilstein Reference: 2375845
MDL No.: MFCD00008529
Formula:
C₂₄H₂₂ MW: 310.44 g/mol Melting Pt: 179…184 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008529 CAS Number: 13280-61-0 EINECS: 236-285-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...