2-sec-Butylthiazole 98+%

Supplier: Thermo Scientific Chemicals
Warning

A13483-06 A13483-14
CAAAA13483-06EA 85.32 CAD
CAAAA13483-06 CAAAA13483-14 CAAAA13483-22
2-sec-Butylthiazole 98+%
2-sec-Butylthiazole

Purity: 98+% CAS No.: 18277-27-5 Molecular Formula: C7H11NS Formula Weight: 141.24 Flash Point: 58°(136°F) Refractive Index: 1.4985 Odor: Characteristic Boiling Point: 174-175° UN No.: 1993 MDL No.: MFCD00051951

Formula: C₇H₁₁NS
Boiling Pt: 174…175 °C
Density: 1.007
Flash Pt: 58 °C (136 °F)
Storage Temperature: Ambient
MDL Number: MFCD00051951
CAS Number: 18277-27-5
EINECS: 242-154-3
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR