Methyl-4-(methylamino)benzoate 98%
Supplier: Thermo Scientific Chemicals
A13460-06
A13460-14
CAAAA13460-06EA
120.87
CAD
CAAAA13460-06
CAAAA13460-14
CAAAA13460-22
Methyl-4-(methylamino)benzoate 98%
Methyl-4-(methylamino)benzoate
Purity: 98%
CAS No.: 18358-63-9
Molecular Formula: C9H11NO2
Formula Weight: 165.19
Melting Point: 92-97°
Beilstein Reference: 2803327
MDL No.: MFCD00017198
Formula:
C₉H₁₁NO₂ Melting Pt: 92…97 °C Storage Temperature: Ambient |
MDL Number:
MFCD00017198 CAS Number: 18358-63-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...